Learn More
Rabbit anti-Goat IgG Fc Secondary Antibody, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen 31133
Additional Details : Weight : 0.80000kg
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product No. 31133 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product No. 31133 reacts with the heavy chains of goat IgG, but not with light chains of most goat immunoglobulins or with non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. After opening, dilute only enough antibody for a single day's use. Store remainder at 4°C under sterile conditions. Country of origin: USA
Thermo Scientific Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Goat IgG Fc | |
Polyclonal | |
Unconjugated | |
Antigen affinity chromatography | |
RUO | |
Goat | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
2.4 mg/mL | |
Rabbit | |
2 mg | |
Secondary | |
4° C | |
IgG |