Learn More
GPC6 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPC6.
Brand: Abnova PAB4470.400uL
Description
The glypicans comprise a family of glycosylphosphatidylinositol-anchored heparan sulfate proteoglycans, and they have been implicated in the control of cell growth and cell division. The glypican encoded by this gene is a putative cell surface coreceptor for growth factors, extracellular matrix proteins, proteases and anti-proteases. [provided by RefSeq
Specifications
| GPC6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GPC6. | |
| In PBS (0.09% sodium azide) | |
| NP_005699;Q9Y625 | |
| GPC6 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human GPC6. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:50-100) The optimal working dilution should be determined by the end user. | |
| GPC6 | |
| MGC126288 | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10082 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only