missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR109B Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPR109B.
Brand: Abnova PAB26280.50ug
This item is not returnable.
View return policy
Specifications
| GPR109B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GPR109B. | |
| In PBS (0.09% sodium azide) | |
| P49019 | |
| GPR109B | |
| A synthetic peptide corresponding to 20 amino acids at C-terminus of human GPR109B. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| GPR109B | |
| HM74/PUMAG/Puma-g | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 8843 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction