missing translation for 'onlineSavingsMsg'
Learn More

GPR124 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16056537
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16056537 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16056537 Supplier Abnova Supplier No. PAB26356.50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of GPR124.

Specifications

Antigen GPR124
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GPR124.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene GPR124
Gene Accession No. Q96PE1
Gene Alias DKFZp434C211/DKFZp434J0911/FLJ14390/KIAA1531/TEM5
Gene Symbols GPR124
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 19 amino acids at N-terminal extracellular domain of human GPR124.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25960
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.