Learn More
GPR132 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPR132.
Brand: Abnova PAB26318.50ug
Description
This gene encodes a subfamily member of the G-protein couple receptor (GPCR) superfamily. The encoded protein is a high-affinity receptor for lysophosphatidylcholine (LPC), a major phospholipid component of oxidized low density lipoprotein. This protein may react to LPC levels at sites of inflammation to limit the expansion of tissue-infiltrating cells. A similar protein in mouse is involved in cell cycle progression. [provided by RefSeq
Specifications
| GPR132 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GPR132. | |
| In PBS (0.09% sodium azide) | |
| Q9UNW8 | |
| GPR132 | |
| A synthetic peptide corresponding to 18 amino acids at 3rd extracellular domain of human GPR132. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins, except PRDM4 (61%). | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-28 ug/mL) The optimal working dilution should be determined by the end user. | |
| GPR132 | |
| G2A/MGC99642 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 29933 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only