Learn More
GPR182 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic protein of GPR182.
Brand: Abnova PAB19566.100ug
Description
Adrenomedullin is a potent vasodilator peptide that exerts major effects on cardiovascular function. This gene encodes a seven-transmembrane protein that belongs to the family 1 of G-protein coupled receptors. Studies of the rat counterpart suggest that the encoded protein may function as a receptor for adrenomedullin. [provided by RefSeq]
Specifications
| GPR182 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic protein of GPR182. | |
| Lyophilized from PBS. | |
| O15218 | |
| GPR182 | |
| Synthetic protein corresponding to human GPR182. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Lyophilized |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| ELISA (1-2 ug/mL) Western Blot (2-10 ug/mL) Immunohistochemistry (2-10 ug/mL) The optimal working dilution should be determined by the end user. | |
| GPR182 | |
| 7TMR/ADMR/AM-R/AMR/G10D/MGC34399/gamrh/hrhAMR | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 11318 | |
| Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only