Learn More
GPRC5B Rabbit anti-Bovine, Dog, Horse, Human, Monkey, Mouse, Rabbit, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPRC5B.
Brand: Abnova PAB26492.50ug
Description
The protein encoded by this gene is a member of the type 3 G protein-coupled receptor family. Members of this superfamily are characterized by a signature 7-transmembrane domain motif. The specific function of this protein is unknown; however, this protein may mediate the cellular effects of retinoic acid on the G protein signal transduction cascade. [provided by RefSeq
Specifications
| GPRC5B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GPRC5B. | |
| In PBS (0.09% sodium azide) | |
| Q9NZH0 | |
| GPRC5B | |
| A synthetic peptide corresponding to 18 amino acids at N-terminal extracellular domain of human GPRC5B. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) The optimal working dilution should be determined by the end user. | |
| GPRC5B | |
| RAIG-2/RAIG2 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 51704 | |
| Bovine, Canine, Equine, Human, Monkey, Mouse, Rabbit, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only