Learn More
GRHL3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GRHL3.
Brand: Abnova PAB15954.100ug
Description
This gene encodes a member of the grainyhead family of transcription factors. The encoded protein interacts with leader-binding protein 32 (LBP-32) and brother of mammalian grainyhead (BOM), and may function as a transcription factor during development. Multiple transcript variants encoding distinct isoforms have been identified for this gene. Additional transcript variants have been described, but their biological nature has not been determined. [provided by RefSeq
Specifications
| GRHL3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GRHL3. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| MGC46624/SOM/TFCP2L4 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| GRHL3 | |
| GRHL3 | |
| A synthetic peptide corresponding to C-terminus of human GRHL3. | |
| RUO | |
| 57822 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only