Learn More
GRK5 Rabbit anti-Bovine, Chicken, Horse, Human, Monkey, Mouse, Pig, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GRK5.
Brand: Abnova PAB16309.50ug
Description
This gene encodes a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. The protein phosphorylates the activated forms of G protein-coupled receptors thus initiating their deactivation. It has also been shown to play a role in regulating the motility of polymorphonuclear leukocytes (PMNs). [provided by RefSeq
Specifications
| GRK5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GRK5. | |
| In PBS (0.09% sodium azide) | |
| P34947 | |
| GRK5 | |
| A synthetic peptide (conjugated with KLH) corresponding to human GRK5. | |
| 50 μg | |
| Primary | |
| Internal domain of human. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (7 ug/mL) The optimal working dilution should be determined by the end user. | |
| GRK5 | |
| GPRK5 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 2869 | |
| Bovine, Chicken, Equine, Human, Monkey, Mouse, Pig, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only