Learn More
GRM2 Rabbit anti-Hamster, Human, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GRM2.
Brand: Abnova PAB26520.50ug
Description
L-glutamate is the major excitatory neurotransmitter in the central nervous system and activates both ionotropic and metabotropic glutamate receptors. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein-coupled receptors, that have been divided into 3 groups on the basis of sequence homology, putative signal transduction mechanisms, and pharmacologic properties. Group I includes GRM1 and GRM5 and these receptors have been shown to activate phospholipase C. Group II includes GRM2 and GRM3 while Group III includes GRM4, GRM6, GRM7 and GRM8. Group II and III receptors are linked to the inhibition of the cyclic AMP cascade but differ in their agonist selectivities. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| GRM2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GRM2. | |
| In PBS (0.09% sodium azide) | |
| Q14416 | |
| GRM2 | |
| A synthetic peptide corresponding to 17 amino acids at N-terminal extracellular domain of human GRM2. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (19 ug/mL) The optimal working dilution should be determined by the end user. | |
| GRM2 | |
| GLUR2/GPRC1B/MGLUR2/mGlu2 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 2912 | |
| Hamster, Human, Rabbit | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only