missing translation for 'onlineSavingsMsg'
Learn More

GUCA2A Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16108910
Change view
Click to view available options
Quantity:
150 μg
Unit Size:
150µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16108910 150 μg 150µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16108910 Supplier Abnova Supplier No. PAB4993.150ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of GUCA2A.

Sequence: PGTCEICAYAACTGC

Specifications

Antigen GUCA2A
Applications ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of GUCA2A.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal)
Gene GUCA2A
Gene Alias GUANYLIN/GUCA2/STARA
Gene Symbols GUCA2A
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human GUCA2A.
Quantity 150 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2980
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.