Learn More
Rabbit anti-Guinea Pig IgG F(ab')2 Secondary Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-72755V
Code produit. 18471457
  • € 336.00 / 0.50mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Guinea Pig samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Spécification
Spécification
IgG F(ab')2
Polyclonal
50mM Sodium Borate
Guinea Pig IgG F(ab')2 fragment
0.5 mL
Adaptive Immunity, Epitope Tags, Immunology
Guinea Pig
Purified
Western Blot, ELISA, Immunohistochemistry
DyLight 405
Rabbit
Immunogen affinity purified
RUO
Secondary
Store at 4°C in the dark.