missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Guinea Pig IgG Fc Secondary Antibody, Janelia Fluor 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-72735JF646
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Guinea Pig samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| IgG Fc | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Guinea Pig IgG F(c) fragment | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| Guinea Pig | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| Janelia Fluor 646 | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction