missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Guinea Pig IgG (H+L) Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-69390
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Guinea Pig samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoblot, Dot Blot
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.2) | |
| Purified normal IgG isolated from pooled guinea pig serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 10 mg | |
| Adaptive Immunity, Immunology | |
| Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoblot, Dot Blot | |
| LYOPH | |
| ELISA 1:100-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000 | |
| Rabbit | |
| Ion exchange chromatography | |
| RUO | |
| Secondary | |
| Guinea Pig | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction