missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Guinea pig IgG whole molecule,Polyclonal Secondary Antibody, Abnova™
Brand: Abnova PAB9253.2mg
This item is not returnable.
View return policy
Specifications
| Rabbit anti-Guinea pig IgG whole molecule | |
| Polyclonal | |
| Rabbit anti-guinea pig IgG recognizes Guinea pig IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. | |
| In 10mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Guinea pig IgG whole molecule | |
| 2 mg | |
| Guinea Pig | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
| Secondary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction