missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H3F3A, H3F3B Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of H3F3A, H3F3B.
Brand: Abnova PAB2226.400uL
This item is not returnable.
View return policy
Specifications
| H3F3A, H3F3B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of H3F3A, H3F3B. | |
| In PBS (0.09% sodium azide) | |
| H3F3A | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 1-30 of human H3 histone. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| Dot Blot, ELISA | |
| Unconjugated | |
| Dot Blot (1:500) The optimal working dilution should be determined by the end user. | |
| H3.3A/H3F3/MGC87782/MGC87783 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 3020/3021 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction