Learn More
HNF1B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant HNF1B.
Brand: Abnova PAB28567.100uL
Description
This gene encodes a member of the homeodomain-containing superfamily of transcription factors. The protein binds to DNA as either a homodimer, or a heterodimer with the related protein hepatocyte nuclear factor 1-alpha. The gene has been shown to function in nephron development, and regulates development of the embryonic pancreas. Mutations in this gene result in renal cysts and diabetes syndrome and noninsulin-dependent diabetes mellitus, and expression of this gene is altered in some types of cancer. Multiple transcript variants encoding different isoforms have been found for this gene
Sequence: KEVLVQALEELLPSPNFGVKLETLPLSPGSGAEPDTKPVFHTLTNGHAKGRLSGDEGSEDGDDYDTPPILKELQALNTEEAAEQRAEVDRMLSEDPWRAAKMIKGYMQQHSpecifications
| HNF1B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant HNF1B. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| FJHN/HNF1beta/HNF2/HPC11/LF-B3/LFB3/MODY5/TCF2/VHNF1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6928 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| Western Blot (1:250-1:500) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:500) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| HNF1B | |
| HNF1B | |
| Recombinant protein corresponding to human HNF1B. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only