Learn More
HNRPDL, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HNRPDL.
Marke: Abnova PAB17744.100ug
Beschreibung
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two RRM domains that bind to RNAs. Two alternatively spliced transcript variants have been described for this gene. One of the variants is probably not translated because the transcript is a candidate for nonsense-mediated mRNA decay. The protein encoded by this gene is similar to its family member HNRPD. [provided by RefSeq
Spezifikation
| HNRPDL | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of HNRPDL. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| HNRNP/JKTBP/JKTBP2/laAUF1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total HNRPDL protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) ELISA (1:20000) The optimal working dilution should be determined by the end user. | |
| HNRPDL | |
| HNRPDL | |
| A synthetic peptide corresponding to internal of human HNRPDL. | |
| RUO | |
| 9987 | |
| Human, Mouse, Rat | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only