Learn More
Rabbit anti-Horse IgG F(c) fragment, FITC, Polyclonal, Secondary Antibody, Abnova™
Brand:  Abnova PAB10673.1500ug
Product Code. 16232980
  • € 406.00 / 1500µg
Specifications
Rabbit anti-Horse IgG F(c) fragment
Polyclonal
Rabbit anti-horse IgG recognizes horse IgG F(c) fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Fluorescein.
Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Horse IgG F(c) fragment
1500 μg
Equine
Lyophilized
Flow Cytometry, Immunofluorescence
FITC
The optimal working dilution should be determined by the end user.
Rabbit
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Secondary
Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.