Learn More
HOXD10, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HOXD10.
Brand: Abnova PAB17750.100ug
Description
This gene is a member of the Abd-B homeobox family and encodes a protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox D genes located on chromosome 2. The encoded nuclear protein functions as a sequence-specific transcription factor that is expressed in the developing limb buds and is involved in differentiation and limb development. Mutations in this gene have been associated with Wilm's tumor and congenital vertical talus (also known as rocker-bottom foot deformity or congenital convex pes valgus) and/or a foot deformity resembling that seen in Charcot-Marie-Tooth disease. [provided by RefSeq]
Specifications
| HOXD10 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of HOXD10. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| HOX4/HOX4D/HOX4E/Hox-4.4 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total HOXD10 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
| HOXD10 | |
| HOXD10 | |
| A synthetic peptide corresponding to internal of human HOXD10. | |
| RUO | |
| 3236 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only