missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HP Rabbit anti-Dog, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native HP.
Brand: Abnova PAB19916.150ug
This item is not returnable.
View return policy
Specifications
| HP | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against native HP. | |
| In PBS, pH 7.5 (0.02% sodium azide, 50% glycerol (v/v)) | |
| HPR | |
| Native purified HP from canine plasma. | |
| 150 μg | |
| Primary | |
| Dog | |
| Liquid |
| ELISA | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| HPR | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 479668 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction