Learn More
HRH3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HRH3.
Brand: Abnova PAB3659.400uL
Description
Histamine is a ubiquitous messenger molecule released from mast cells, enterochromaffin-like cells, and neurons. Its various actions are mediated by histamine receptors H1, H2, H3 and H4. This gene encodes one of the histamine receptors (H3) which belongs to the family 1 of G protein-coupled receptors. It is an integral membrane protein and can regulate neurotransmitter release. This receptor can also increase voltage-dependent calcium current in smooth muscles and innervates the blood vessels and the heart in cardiovascular system. [provided by RefSeq
Specifications
| HRH3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of HRH3. | |
| In PBS (0.09% sodium azide) | |
| GPCR97/HH3R | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 11255 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:10-50) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| HRH3 | |
| HRH3 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human HRH3. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only