Learn More
HSF1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HSF1.
Brand: Abnova PAB4339.400uL
Description
The product of this gene is a heat-shock transcription factor. Transcription of heat-shock genes is rapidly induced after temperature stress. Hsp90, by itself and/or associated with multichaperone complexes, is a major repressor of this gene. [provided by RefSeq
Sequence: PLVRVKEEPPSPPQSPRSpecifications
| HSF1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of HSF1. | |
| In PBS (0.09% sodium azide) | |
| HSTF1 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 3297 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
| HSF1 | |
| HSF1 | |
| A synthetic peptide (conjugated with KLH) corresponding to human HSF1. | |
| 400 μL | |
| Primary | |
| This antibody recognize synthetic peptide (PLVRVKEEPPSPPQSPR) containing a predicted sumoylation site of human HSF1. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only