missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90 Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Hsp90.
Brand: Abnova PAB14473.100uL
This item is not returnable.
View return policy
Specifications
| HSP90 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against Hsp90. | |
| In serum | |
| Full length Hsp90 protein. | |
| RUO | |
| This antibody recognizes eukaryotic Hsp90alpha/beta. | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:20000-1:40000) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction