missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90B1, Rabbit, ATTO488, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Hsp90b1.
Brand: Abnova PAB28948.100ug
This item is not returnable.
View return policy
Specifications
| HSP90B1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Hsp90b1. | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| ERp99/GRP94/TA-3/Targ2/Tra-1/Tra1/gp96 | |
| Rabbit | |
| Peptide Affinity Purified | |
| RUO | |
| 22027 | |
| Bovine, Human, Mouse, Rat | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Atto 488 | |
| Immunocytochemistry Immunohistochemistry Immunoprecipitation (1:80) Western blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Hsp90b1 | |
| Hsp90b1 | |
| A synthetic peptide corresponding to amino acids at C-terminus of mouse Hsp90b1. | |
| 100 μg | |
| Primary | |
| Detects ∼94 kDa. | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction