Learn More
HSPA5 Rabbit anti-Bovine, Chicken, Fruit fly, Human, Mouse, Nematoda, Primates, Rat, Sheep, Zebra fish, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HSPA5.
Brand: Abnova PAB12413.100uL
Description
When Chinese hamster K12 cells are starved of glucose, the synthesis of several proteins, called glucose-regulated proteins (GRPs), is markedly increased. Hendershot et al. (1994) [PubMed 8020977] pointed out that one of these, GRP78 (HSPA5), also referred to as 'immunoglobulin heavy chain-binding protein' (BiP), is a member of the heat-shock protein-70 (HSP70) family and is involved in the folding and assembly of proteins in the endoplasmic reticulum (ER). Because so many ER proteins interact transiently with GRP78, it may play a key role in monitoring protein transport through the cell.[supplied by OMIM
Specifications
| HSPA5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of HSPA5. | |
| In PBS (30% glycerol, 0.09% sodium azide) | |
| BIP/FLJ26106/GRP78/MIF2 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Bovine, Chicken, Drosophila, Human, Mouse, Nematode, Primate, Rat, Sheep, Zebrafish | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| HSPA5 | |
| HSPA5 | |
| A synthetic peptide corresponding to amino acids 250-300 of human HSPA5. | |
| RUO | |
| 3309 | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only