Learn More
HSPB1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HSPB1.
Brand: Abnova PAB2048.400uL
Description
The protein encoded by this gene is induced by environmental stress and developmental changes. The encoded protein is involved in stress resistance and actin organization and translocates from the cytoplasm to the nucleus upon stress induction. Defects in this gene are a cause of Charcot-Marie-Tooth disease type 2F (CMT2F) and distal hereditary motor neuropathy (dHMN). [provided by RefSeq
Specifications
| HSPB1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of HSPB1. | |
| In PBS (0.09% sodium azide) | |
| CMT2F/DKFZp586P1322/HMN2B/HS.76067/HSP27/HSP28/Hsp25/SRP27 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 3315 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. | |
| HSPB1 | |
| HSPB1 | |
| A synthetic peptide (conjugated with KLH) corresponding to residues surrounding S82 of human HSPB1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only