Learn More
HSPB1 (phospho S78), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of HSPB1.
Brand: Abnova PAB24728.100uL
Description
The protein encoded by this gene is induced by environmental stress and developmental changes. The encoded protein is involved in stress resistance and actin organization and translocates from the cytoplasm to the nucleus upon stress induction. Defects in this gene are a cause of Charcot-Marie-Tooth disease type 2F (CMT2F) and distal hereditary motor neuropathy (dHMN). [provided by RefSeq
Specifications
| HSPB1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of HSPB1. | |
| In PBS, pH 7.2 (0.09% sodium azide) | |
| CMT2F/DKFZp586P1322/HMN2B/HS.76067/HSP27/HSP28/Hsp25/SRP27 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3315 | |
| Human, Mouse, Rat | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| HSPB1 | |
| HSPB1 | |
| Synthetic phosphopeptide corresponding to residues surrounding S78 of HSPB1. | |
| 100 μL | |
| Primary | |
| HSPB1 (phospho S78) polyclonal antibody detects endogenous levels of HSPB1 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only