Learn More
HTR4 Rabbit anti-Bovine, Dog, Horse, Human, Monkey, Pig, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HTR4.
Brand: Abnova PAB25559.50ug
Description
This gene is a member of the family of serotonin receptors, which are G protein coupled receptors that stimulate cAMP production in response to serotonin (5-hydroxytryptamine). The gene product is a glycosylated transmembrane protein that functions in both the peripheral and central nervous system to modulate the release of various neurotransmitters. Multiple transcript variants encoding proteins with distinct C-terminal sequences have been described, but the full-length nature of some transcript variants has not been determined. [provided by RefSeq
Specifications
| HTR4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of HTR4. | |
| In PBS (0.09% sodium azide) | |
| 5-HT4/5-HT4R | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 3360 | |
| Bovine, Canine, Equine, Human, Monkey, Pig, Rabbit | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| HTR4 | |
| HTR4 | |
| A synthetic peptide corresponding to 15 amino acids at 3rd cytoplasmic domain of human HTR4. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only