Learn More
Rabbit anti-Human IgG F(ab')2 Secondary Antibody, Janelia Fluor 549 (Pre-adsorbed), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-72785JF549
This item is not returnable.
View return policy
Description
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| IgG F(ab')2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit anti-Human IgG F(ab)2 Secondary Antibody (Pre-adsorbed)was produced in rabbit by repeated immunization with human IgG F(ab')2 fragment. | |
| 0.5 mL | |
| Adaptive Immunity, Epitope Tags, Immunology | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| Janelia Fluor 549 | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. |