missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Human IgG Heavy Chain Secondary Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22138H
This item is not returnable.
View return policy
Description
IgG Heavy Chain Polyclonal antibody specifically detects IgG Heavy Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot
Specifications
| IgG Heavy Chain | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Dot Blot | |
| HRP | |
| Immunoglobulin G, ImmunoglobulinG | |
| Human IgG gamma heavy chain | |
| 0.5 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction