missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Human IgG1 Fc Secondary Antibody (245), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-40578APC
This item is not returnable.
View return policy
Description
IgG1 Fc Monoclonal antibody specifically detects IgG1 Fc in Human samples. It is validated for ELISA
Specifications
| IgG1 Fc | |
| Monoclonal | |
| APC | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
| ELISA | |
| 245 | |
| PBS | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IgG1 Fc (Uniprot#: P01857-1; Glu99-Lys330,C103S). HEK293 used as expression system. | |
| 0.1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction