missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aspergillus, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Aspergillus.
Brand: Abnova PAB13893.1mL
This item is not returnable.
View return policy
Specifications
| Aspergillus | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against Aspergillus. | |
| In 10mM PBS, pH 7.2 (0.09% sodium azide) | |
| Soluble extract from Aspergillus fumigatus, Aspergillus flavus, Aspergillus niger and Aspergillus terreus. | |
| RUO | |
| Reactive with soluble proteins from common Aspergillus species. Antiserum is not absorbed and may react with related microorganisms. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 1 mL | |
| Primary | |
| Bacteria, Fungi | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction