missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bacillus spores, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Bacillus spores.
Brand: Abnova PAB13894.1mL
This item is not returnable.
View return policy
Specifications
| Bacillus spores | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against Bacillus spores. | |
| In 10mM PBS, pH 7.2 (0.09% sodium azide) | |
| Native purified fromBacillus cereus (ATCC #11778) and Bacillus subtilis (ATCC #9372). | |
| RUO | |
| Reactive with spores and vegetative cells of Bacillus cereus and Bacillus subtilis. Antiserum is unabsorbed and may cross-react with other Bacillus species. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 1 mL | |
| Primary | |
| Bacteria | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction