missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlamydia trachomatis (Elementary Bodies), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Chlamydia trachomatis (Elementary Bodies).
Brand: Abnova PAB13906.1mL
This item is not returnable.
View return policy
Specifications
| Chlamydia trachomatis (Elementary Bodies) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against Chlamydia trachomatis (Elementary Bodies). | |
| In 10mM PBS, pH 7.2 (0.09% sodium azide) | |
| L2 and other serovar groups. | |
| RUO | |
| Recognizes purified elementary bodies, disrupted (all serovars A-K, L1-L3). Cross-reacts with Chlamydia psittacii and Chlamydia pneumoniae (TWAR). Negative uninfected cell reactivity against HEp-2 cells and egg yolk sac. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 1 mL | |
| Primary | |
| Chlamydomonas species | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction