missing translation for 'onlineSavingsMsg'
Learn More

Escherichia coli (O/K antigens), Rabbit, Polyclonal Antibody, Abnova™

Product Code. 16001007
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16001007 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16001007 Supplier Abnova Supplier No. PAB13941.1mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against Escherichia coli (O/K antigens).

Specifications

Antigen Escherichia coli (O/K antigens)
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Escherichia coli (O/K antigens).
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Rabbit
Immunogen Mixture of Escherichia coli serotypes.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Many “O” and “K” antigenic serotypes of Escherichia coli. Antiserum is not absorbed and does cross-react with related Enterobacteriaceae. Will remove E. coli proteins from recombinant preparations.
Target Species E. coli
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.