missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Helicobacter pylori Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Helicobacter pylori.
Brand: Abnova PAB13961.1mL
This item is not returnable.
View return policy
Specifications
| Helicobacter pylori | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against Helicobacter pylori. | |
| In 10mM PBS, pH 7.2 (0.09% sodium azide) | |
| Native from Helicobacter pylori whole organism (ATCC strain 43504). | |
| RUO | |
| Whole cell lysate. Has not been absorbed and may react with related microorganisms. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 1 mL | |
| Primary | |
| H. pylori | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction