missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pseudomonas aeruginosa Tail Fiber Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Pseudomonas aeruginosa Tail Fiber.
Brand: Abnova PAB16039.100ug
This item is not returnable.
View return policy
Specifications
| Pseudomonas aeruginosa Tail Fiber | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Pseudomonas aeruginosa Tail Fiber. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| A synthetic peptide corresponding to N-terminus of Pseudomonas aeruginosa Tail Fiber. | |
| RUO | |
| P. aeruginosa | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction