missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salmonella, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Salmonella.
Brand: Abnova PAB13824.1mL
This item is not returnable.
View return policy
Specifications
| Salmonella | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against Salmonella. | |
| In 10mM PBS, pH 7.2 (0.09% sodium azide) | |
| A mixture of Salmonella enteriditis, Salmonella typhimurium, and Salmonella heidelburg. | |
| RUO | |
| Polyvalent for Salmonella “O” & “H” antigens. Immunocaptures Salmonellae. Antiserum is not absorbed and does react with related Enterobacteriaceae. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 1 mL | |
| Primary | |
| Salmonella | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction