missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Schmidtea mediterranea Dvl-b, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Schmidtea mediterranea Dvl-b.
Brand: Abnova PAB16048.100ug
This item is not returnable.
View return policy
Specifications
| Schmidtea mediterranea Dvl-b | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Schmidtea mediterranea Dvl-b. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| A synthetic peptide corresponding to C-terminus of Freshwater Flatworms Schmidtea mediterranea Dvl-b. | |
| RUO | |
| Parasite | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction