missing translation for 'onlineSavingsMsg'
Learn More

IFI27 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16031368
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16031368 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16031368 Supplier Abnova Supplier No. PAB8961.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of IFI27.

Specifications

Antigen IFI27
Applications Dot Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of IFI27.
Dilution ELISA (1:10000-1:100000) Dot Blot (1:10000-1:100000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene IFI27
Gene Alias FAM14D/ISG12/ISG12A/P27
Gene Symbols IFI27
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus human IFI27.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3429
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.