missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgA Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native IgA.
Brand: Abnova PAB16873.500ug
This item is not returnable.
View return policy
Specifications
| IgA | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against native IgA. | |
| In 10mM PBS (50% glycerol, 0.01% BSA, 0.05% sodium azide) | |
| Native purified IgA from human. | |
| RUO | |
| This antibody mainly recognizes human IgA heavy chain. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:80000) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 500 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction