Learn More
IGF1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant IGF1.
Brand: Abnova PAB19852.150ug
Description
The protein encoded by this gene is similar to insulin in function and structure and is a member of a family of proteins involved in mediating growth and development. The encoded protein is processed from a precursor, bound by a specific receptor, and secreted. Defects in this gene are a cause of insulin-like growth factor I deficiency. Several transcript variants encoding different isoforms have been found for this gene
Specifications
| IGF1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant IGF1. | |
| In PBS, pH 7.5 (0.02% sodium azide, 50% glycerol (v/v)) | |
| IGFI | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 3479 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| IGF1 | |
| IGF1 | |
| Recombinant protein corresponding to human IGF1. | |
| 150 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only