Learn More
IL16 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of IL16.
Brand: Abnova PAB16687.100ug
Description
The protein encoded by this gene is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of this gene undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| IL16 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of IL16. | |
| In PBS (0.02% sodium azide) | |
| FLJ16806/FLJ42735/FLJ44234/HsT19289/IL-16/LCF/prIL-16 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| IL16 | |
| IL16 | |
| A synthetic peptide corresponding to N-terminus 20 amino acids of human IL16. | |
| RUO | |
| 3603 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only