missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL17F Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against full-length recombinant Il17f.
Brand: Abnova PAB16929.100ug
This item is not returnable.
View return policy
Specifications
| IL17F | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against full length recombinant Il17f. | |
| Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 | |
| C87042/IL-17F | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Lyophilized |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Il17f | |
| Il17f | |
| Recombinant protein corresponding to full length mouse Il17f. | |
| RUO | |
| 257630 | |
| Store at 4°C on dry atmosphere. After reconstitution with 0.1mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction