missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1F5, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against full-length recombinant IL1F5.
Brand: Abnova PAB16917.100ug
This item is not returnable.
View return policy
Specifications
| IL1F5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against full length recombinant IL1F5. | |
| Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Recombinant protein corresponding to 155 amino acids of bovine IL1F5. | |
| RUO | |
| Bovine | |
| Lyophilized |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:8000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Store at 4°C on dry atmosphere. After reconstitution with 0.1mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction