Learn More
IL1RL1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant IL1RL1.
Brand: Abnova PAB20396.100uL
Description
The protein encoded by this gene is a member of the interleukin 1 receptor family. Studies of the similar gene in mouse suggested that this receptor can be induced by proinflammatory stimuli, and may be involved in the function of helper T cells. This gene, interleukin 1 receptor, type I (IL1R1), interleukin 1 receptor, type II (IL1R2) and interleukin 1 receptor-like 2 (IL1RL2) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq
Sequence: PSYTVDWYYSQTNKSIPTQERNRVFASGQLLKFLPAAVADSGIYTCIVRSPTFNRTGYANVTIYKKQSDCNVPDYLMYSTVSGSEKNSKIYCPTIDLYNWTAPLEWFKNCQALQGSRYSpecifications
| IL1RL1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant IL1RL1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DER4/FIT-1/MGC32623/ST2/ST2L/ST2V/T1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9173 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| IL1RL1 | |
| IL1RL1 | |
| Recombinant protein corresponding to amino acids of human IL1RL1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only