missing translation for 'onlineSavingsMsg'
Learn More

IL21 Rabbit anti-Bovine, Horse, Pig, Polyclonal Antibody, Abnova™

Product Code. 16042467
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16042467 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16042467 Supplier Abnova Supplier No. PAB16898.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against full-length recombinant IL21.

Specifications

Antigen IL21
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant IL21.
Dilution ELISA (1:6000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 24-152 of bovine IL21.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine, Equine, Pig
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.