Learn More
INHA Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of INHA.
Marke: Abnova PAB8965.100ug
Beschreibung
The inhibin alpha subunit joins either the beta A or beta B subunit to form a pituitary FSH secretion inhibitor. Inhibin has been shown to regulate gonadal stromal cell proliferation negatively and to have tumour-suppressor activity. In addition, serum levels of inhibin have been shown to reflect the size of granulosa-cell tumors and can therefore be used as a marker for primary as well as recurrent disease. However, in prostate cancer, expression of the inhibin alpha-subunit gene was suppressed and was not detectable in poorly differentiated tumor cells. Furthermore, because expression in gonadal and various extragonadal tissues may vary severalfold in a tissue-specific fashion, it is proposed that inhibin may be both a growth/differentiation factor and a hormone. [provided by RefSeq
Spezifikation
| INHA | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of INHA. | |
| In buffer containing 0.02% sodium azide | |
| INHA | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 18-23 of human INHA. | |
| RUO | |
| 3623 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Dot Blot, ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| ELISA (1:20000) Dot Blot (1:20000) Western Blot (1:500) Immunohistochemistry (1:200) The optimal working dilution should be determined by the end user. | |
| INHA | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only