missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF4 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of IRF4.
Brand: Abnova PAB26943.100uL
This item is not returnable.
View return policy
Specifications
| IRF4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of IRF4. | |
| In PBS, pH 7.2 (0.09% sodium azide) | |
| LSIRF/MUM1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 3662 | |
| Human, Mouse | |
| Liquid |
| Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| IRF4 | |
| IRF4 | |
| A synthetic peptide corresponding to human IRF4. | |
| 100 μL | |
| Primary | |
| IRF4 polyclonal antibody detects endogenous levels of IRF4 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction